missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 1-Methyl-3-piperidinecarboxylate, TCI America™

Manufacturer:  TCI America E086325G

 View more versions of this product

Catalog No. E086325G



Specifications

Specifications

Ethyl 1-Methyl-3-piperidinecarboxylate
Yellow
5166-67-6
MFCD00006496
VFJJNMLPRDRTCO-UHFFFAOYSA-N
ethyl 1-methylpiperidine-3-carboxylate
97981
Clear Liquid at 20°C
96°C
25g
C9H17NO2
ethyl 1-methylnipecotate, ethyl n-methylpiperidine-3-carboxylate, ethyl 1-methyl-3-piperidinecarboxylate, 1-methyl-piperidine-3-carboxylic acid ethyl ester, 1-methylpiperidine-3-carboxylic acid ethyl ester, 1-methyl-3-piperidinecarboxylic acid ethyl ester, 3-piperidinecarboxylic acid, 1-methyl-, ethyl ester, ethyl-1-methylpiperidin-3-carboxylat, pubchem11774, acmc-209kvb
CCOC(=O)C1CCCN(C1)C
171.24
171.24
≥95.0% (GC,T)

Chemical Identifiers

5166-67-6
171.24
VFJJNMLPRDRTCO-UHFFFAOYSA-N
97981
CCOC(=O)C1CCCN(C1)C
C9H17NO2
MFCD00006496
ethyl 1-methylnipecotate, ethyl n-methylpiperidine-3-carboxylate, ethyl 1-methyl-3-piperidinecarboxylate, 1-methyl-piperidine-3-carboxylic acid ethyl ester, 1-methylpiperidine-3-carboxylic acid ethyl ester, 1-methyl-3-piperidinecarboxylic acid ethyl ester, 3-piperidinecarboxylic acid, 1-methyl-, ethyl ester, ethyl-1-methylpiperidin-3-carboxylat, pubchem11774, acmc-209kvb
ethyl 1-methylpiperidine-3-carboxylate
Safety and Handling

Safety and Handling

TSCA : No

Documents