missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 1-methyl-1H-benzimidazole-5-carboxylate, 97%, Maybridge

Chemical Identifiers

CAS 53484-19-8
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C11H12N2O2
Formula Weight 204.23
MDL Number MFCD04501518
Chemical Name or Material Ethyl 1-methyl-1H-benzimidazole-5-carboxylate
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC13623DE SDS Fisher Scientific Company
CC13623DE
Amber Glass Bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

53484-19-8
100.0
C11H12N2O2
MFCD04501518
97%
96.5
97%
204.23
Ethyl 1-methyl-1H-benzimidazole-5-carboxylate

Specifications

204.23
100.0
C11H12N2O2
MFCD04501518
Amber Glass Bottle
97%
5g
97%
53484-19-8
96.5
Ethyl 1-methyl-1H-benzimidazole-5-carboxylate

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning