missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 1-(hydroxymethyl)cyclohexanecarboxylate, 97%

$83.14 - $1,207.49

Chemical Identifiers

CAS 834914-39-5
Molecular Formula C10H18O3
Molecular Weight (g/mol) 186.251
MDL Number MFCD12022627
InChI Key CHMBRLRRLJHXRK-UHFFFAOYSA-N
Synonym ethyl 1-hydroxymethyl cyclohexanecarboxylate, cyclohexanecarboxylic acid, 1-hydroxymethyl-, ethyl ester, ethyl 1-hydroxymethyl cyclohexane-1-carboxylate, ethyl 1-hydroxymethyl cyclohexane carboxylate, 1-hydroxymethyl-cyclohexanecarboxylic acid ethyl ester
PubChem CID 46172882
IUPAC Name ethyl 1-(hydroxymethyl)cyclohexane-1-carboxylate
SMILES CCOC(=O)C1(CCCCC1)CO
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62519MD View Documents Alfa Aesar™
H62519MD
250mg Each for $83.14
 
AAH6251903 View Documents Alfa Aesar™
H6251903
1g Each for $263.70
 
AAH6251906 View Documents Alfa Aesar™
H6251906
5g Each for $1,207.49
 
Specifications

Chemical Identifiers

834914-39-5
186.251
CHMBRLRRLJHXRK-UHFFFAOYSA-N
46172882
CCOC(=O)C1(CCCCC1)CO
C10H18O3
MFCD12022627
ethyl 1-hydroxymethyl cyclohexanecarboxylate, cyclohexanecarboxylic acid, 1-hydroxymethyl-, ethyl ester, ethyl 1-hydroxymethyl cyclohexane-1-carboxylate, ethyl 1-hydroxymethyl cyclohexane carboxylate, 1-hydroxymethyl-cyclohexanecarboxylic acid ethyl ester
ethyl 1-(hydroxymethyl)cyclohexane-1-carboxylate

Specifications

250mg
C10H18O3
ethyl 1-hydroxymethyl cyclohexanecarboxylate, cyclohexanecarboxylic acid, 1-hydroxymethyl-, ethyl ester, ethyl 1-hydroxymethyl cyclohexane-1-carboxylate, ethyl 1-hydroxymethyl cyclohexane carboxylate, 1-hydroxymethyl-cyclohexanecarboxylic acid ethyl ester
CCOC(=O)C1(CCCCC1)CO
186.251
186.24
Ethyl 1-(hydroxymethyl)cyclohexanecarboxylate
834914-39-5
MFCD12022627
CHMBRLRRLJHXRK-UHFFFAOYSA-N
ethyl 1-(hydroxymethyl)cyclohexane-1-carboxylate
46172882
97%