missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 1-(hydroxymethyl)cyclohexanecarboxylate, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6251903
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Ethyl 1-(hydroxymethyl)cyclohexanecarboxylate | |
C10H18O3 | |
1 g | |
CHMBRLRRLJHXRK-UHFFFAOYSA-N | |
ethyl 1-(hydroxymethyl)cyclohexane-1-carboxylate | |
46172882 | |
97% |
834914-39-5 | |
MFCD12022627 | |
ethyl 1-hydroxymethyl cyclohexanecarboxylate, cyclohexanecarboxylic acid, 1-hydroxymethyl-, ethyl ester, ethyl 1-hydroxymethyl cyclohexane-1-carboxylate, ethyl 1-hydroxymethyl cyclohexane carboxylate, 1-hydroxymethyl-cyclohexanecarboxylic acid ethyl ester | |
CCOC(=O)C1(CCCCC1)CO | |
186.251 | |
186.24 |
Chemical Identifiers
834914-39-5 | |
186.251 | |
CHMBRLRRLJHXRK-UHFFFAOYSA-N | |
46172882 | |
CCOC(=O)C1(CCCCC1)CO |
C10H18O3 | |
MFCD12022627 | |
ethyl 1-hydroxymethyl cyclohexanecarboxylate, cyclohexanecarboxylic acid, 1-hydroxymethyl-, ethyl ester, ethyl 1-hydroxymethyl cyclohexane-1-carboxylate, ethyl 1-hydroxymethyl cyclohexane carboxylate, 1-hydroxymethyl-cyclohexanecarboxylic acid ethyl ester | |
ethyl 1-(hydroxymethyl)cyclohexane-1-carboxylate |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures