Please login to your online account to display your discounted pricing

Ethyl 1-hydroxycyclopropanecarboxylate, 90%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  335300010

 View more versions of this product

Catalog No. AC335300010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥88%
Formula Weight 130.14
Physical Form Liquid
Quantity 1g
Color Clear light yellow to brown
Refractive Index 1.4400 to 1.4470 (20°C, 589nm)
Packaging Glass bottle
Infrared Spectrum Authentic
CAS 137682-89-4
Chemical Name or Material Ethyl 1-hydroxycyclopropanecarboxylate
Linear Formula C3H4(OH)COOCHCH3
MDL Number MFCD01863257
Name Note 90%
Molecular Formula C6H10O3
Assay Percent Range 90%

Specifications

Chemical Name or Material Ethyl 1-hydroxycyclopropanecarboxylate
Name Note 90%
CAS 137682-89-4
Assay ≥88%
Assay Percent Range 90%
Molecular Formula C6H10O3
Linear Formula C3H4(OH)COOCHCH3
Formula Weight 130.14
MDL Number MFCD01863257