Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl(1-hexadecyl)dimethylammonium bromide, 99%

$41.14 - $278.76

Chemical Identifiers

Chemical Name or Material Ethyl(1-hexadecyl)dimethylammonium bromide
CAS 124-03-8
Molecular Formula CH3(CH2)15N(CH3)2(CH2CH3)Br
Formula Weight 378.49
MDL Number MFCD00011774
Beilstein 3729134
Merck Index 14,2029
Synonym Cetyl ethyl dimethylammonium bromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2399114

  SDS
alfa aesar™
B2399114
25g Each for $41.14

AAB2399122

  SDS
alfa aesar™
B2399122
100g Each for $95.99

AAB2399136

  SDS
alfa aesar™
B2399136
500g Each for $278.76
Description & Specifications

Specifications

Formula Weight 378.49
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number BQ5425000
Melting Point ≈192°C (dec.)
Synonym Cetyl ethyl dimethylammonium bromide
Molecular Formula CH3(CH2)15N(CH3)2(CH2CH3)Br
Beilstein 3729134
Merck Index 14,2029
Assay 99%
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 25g
TSCA TSCA
CAS 124-03-8
Chemical Name or Material Ethyl(1-hexadecyl)dimethylammonium bromide
MDL Number MFCD00011774
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.