Learn More
Ethyl 1-bromocyclobutanecarboxylate, 96%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 188710010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Ethyl 1-bromocyclobutanecarboxylate | |
1.2790g/mL | |
79°C | |
95% min. (GC) | |
C7H11BrO2 | |
MFCD00001322 | |
1.279 | |
UTVNSHXHFRIXMM-UHFFFAOYSA-N | |
ethyl 1-bromocyclobutane-1-carboxylate | |
97595 | |
96% |
35120-18-4 | |
85°C to 88°C (12.0mmHg) | |
Authentic | |
Glass bottle | |
BrC4H6CO2C2H5 | |
1g | |
ethyl 1-bromocyclobutanecarboxylate, ethyl alpha-bromocyclobutanecarboxylate, 1-bromo-cyclobutanecarboxylic acid ethyl ester, cyclobutanecarboxylic acid, 1-bromo-, ethyl ester, 1-bromocyclobutanecarboxylic acid ethyl ester, ethyl .alpha.-bromocyclobutanecarboxylate, acmc-1cswe, ethyl abromocyclobutanecarboxylate, ethyl-1-bromocyclobutanecarboxylate, 1-ethoxycarbonyl-1-bromocyclobutane | |
CCOC(=O)C1(CCC1)Br | |
207.06 | |
207.06 |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
If eye irritation persists: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
EINECSNumber : 252-384-6
RUO â Research Use Only