Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 1-Boc-piperazine-3-carboxylate, 97%

Chemical Identifiers

Chemical Name or Material Ethyl 1-Boc-piperazine-3-carboxylate
CAS 183742-29-2
Formula Weight 258.31
MDL Number MFCD13172266
Synonym 1-Boc-piperazine-3-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6314603

 
alfa aesar™
H6314603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6314606

 
alfa aesar™
H6314606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63146MD

 
alfa aesar™
H63146MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 258.31
CAS 183742-29-2
Quantity 1g
Chemical Name or Material Ethyl 1-Boc-piperazine-3-carboxylate
Assay 97%
Synonym 1-Boc-piperazine-3-carboxylic acid ethyl ester
MDL Number MFCD13172266
Physical Form Solid