missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 1-Benzylpyrrole-3-carboxylate, TCI America™

$171.11 - $522.52

Chemical Identifiers

CAS 128259-47-2
Molecular Formula C14H15NO2
Molecular Weight (g/mol) 229.279
MDL Number MFCD06797056
InChI Key KSZAVNAKDOXZGJ-UHFFFAOYSA-N
Synonym 1-Benzylpyrrole-3-carboxylic Acid Ethyl Ester
PubChem CID 14667729
IUPAC Name ethyl 1-benzylpyrrole-3-carboxylate
SMILES CCOC(=O)C1=CN(C=C1)CC2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B22841G View Documents TCI America
B22841G
1g Each for $171.11
 
B22845G View Documents TCI America
B22845G
5g Each for $522.52
 
Specifications

Chemical Identifiers

128259-47-2
229.279
KSZAVNAKDOXZGJ-UHFFFAOYSA-N
14667729
CCOC(=O)C1=CN(C=C1)CC2=CC=CC=C2
C14H15NO2
MFCD06797056
1-Benzylpyrrole-3-carboxylic Acid Ethyl Ester
ethyl 1-benzylpyrrole-3-carboxylate

Specifications

Yellow
128259-47-2
MFCD06797056
KSZAVNAKDOXZGJ-UHFFFAOYSA-N
ethyl 1-benzylpyrrole-3-carboxylate
14667729
Clear Liquid at 20°C
Ethyl 1-Benzylpyrrole-3-carboxylate
1g
C14H15NO2
1-Benzylpyrrole-3-carboxylic Acid Ethyl Ester
CCOC(=O)C1=CN(C=C1)CC2=CC=CC=C2
229.279
229.28
≥95.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator