missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 1-Benzyl-4-piperidinecarboxylate, TCI America™

Manufacturer:  TCI America E08625G

 View more versions of this product

Catalog No. E08625G



Specifications

Specifications

Ethyl 1-Benzyl-4-piperidinecarboxylate
Red
24228-40-8
MFCD00040748
ASQCOPJFYLJCGD-UHFFFAOYSA-N
ethyl 1-benzylpiperidine-4-carboxylate
90423
Clear Liquid at 20°C
136°C
5g
C15H21NO2
ethyl n-benzylpiperidine-4-carboxylate, 1-benzylpiperidine-4-carboxylic acid ethyl ester, 1-benzyl-piperidine-4-carboxylic acid ethyl ester, ethyl 1-benzyl-4-piperidinecarboxylate, ethyl 1-benzylisonipecotate, 4-piperidinecarboxylic acid, 1-phenylmethyl-, ethyl ester, 1-benzyl-4-piperidinecarboxylic acid ethyl ester, ethyl n-benzyl-isonipecotate, 1-benzyl-4-carboethoxypiperidine, 1-benzyl-piperidine-4-carboxylic acid-ethyl ester
CCOC(=O)C1CCN(CC1)CC2=CC=CC=C2
247.338
247.34
≥95.0% (GC,T)

Chemical Identifiers

24228-40-8
247.338
ASQCOPJFYLJCGD-UHFFFAOYSA-N
90423
CCOC(=O)C1CCN(CC1)CC2=CC=CC=C2
C15H21NO2
MFCD00040748
ethyl n-benzylpiperidine-4-carboxylate, 1-benzylpiperidine-4-carboxylic acid ethyl ester, 1-benzyl-piperidine-4-carboxylic acid ethyl ester, ethyl 1-benzyl-4-piperidinecarboxylate, ethyl 1-benzylisonipecotate, 4-piperidinecarboxylic acid, 1-phenylmethyl-, ethyl ester, 1-benzyl-4-piperidinecarboxylic acid ethyl ester, ethyl n-benzyl-isonipecotate, 1-benzyl-4-carboethoxypiperidine, 1-benzyl-piperidine-4-carboxylic acid-ethyl ester
ethyl 1-benzylpiperidine-4-carboxylate
Safety and Handling

Safety and Handling

TSCA : No

Documents