missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 1-Aminocyclopropanecarboxylate Hydrochloride, TCI America™

Manufacturer:  TCI America E09375G

 View more versions of this product

Catalog No. E09375G



Specifications

Specifications

Ethyl 1-Aminocyclopropanecarboxylate Hydrochloride
110°C
42303-42-4
MFCD00190747
XFNUTZWASODOQK-UHFFFAOYSA-N
ethyl 1-aminocyclopropane-1-carboxylate;hydrochloride
386203
Crystal-Powder at 20°C
White
5g
C6H12ClNO2
ethyl 1-aminocyclopropanecarboxylate hydrochloride, 1-aminocyclopropane-1-carboxylic acid ethyl ester hydrochloride, h-acpc-oet.hcl, ethyl 1-aminocyclopropane-1-carboxylate hydrochloride, 1-amino-cyclopropyl-1-carboxylic acid ethyl ester hydrochloride, h-acpc-oet hcl, 1-aminocyclopropane-1-carboxylic acid ethyl ester, acpc-oet-hcl, h-acpc-oet?cl, pubchem14733
CCOC(=O)C1(CC1)N.Cl
165.617
165.62
≥98.0% (N,T)

Chemical Identifiers

42303-42-4
165.617
XFNUTZWASODOQK-UHFFFAOYSA-N
386203
CCOC(=O)C1(CC1)N.Cl
C6H12ClNO2
MFCD00190747
ethyl 1-aminocyclopropanecarboxylate hydrochloride, 1-aminocyclopropane-1-carboxylic acid ethyl ester hydrochloride, h-acpc-oet.hcl, ethyl 1-aminocyclopropane-1-carboxylate hydrochloride, 1-amino-cyclopropyl-1-carboxylic acid ethyl ester hydrochloride, h-acpc-oet hcl, 1-aminocyclopropane-1-carboxylic acid ethyl ester, acpc-oet-hcl, h-acpc-oet?cl, pubchem14733
ethyl 1-aminocyclopropane-1-carboxylate;hydrochloride
Safety and Handling

Safety and Handling

TSCA : No

Documents