Please login to your online account to display your discounted pricing

Ethyl 1,3-dithiolane-2-carboxylate, 99%, ACROS Organics™


Quantity 25mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  209990250

 View more versions of this product

Catalog No. AC209990250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 178.26
Physical Form Liquid
Quantity 25mL
Color Clear light yellow
Beilstein 19, II, 290
EINECS Number 243-837-9
Density 1.24g/mL
Refractive Index 1.5380 to 1.5400 (20°C, 89nm)
Infrared Spectrum Authentic
Packaging Glass bottle
Synonym Glyoxylic acid ethyl ester ethylene mercaptal
Name Note 99%
MDL Number MFCD00005411
Assay Percent Range 99%
Molecular Formula C6H10O2S2
Chemical Name or Material Ethyl 1, 3-dithiolane-2-carboxylate
CAS 20461-99-8

Specifications

Chemical Name or Material Ethyl 1, 3-dithiolane-2-carboxylate
Name Note 99%
CAS 20461-99-8
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H10O2S2
Formula Weight 178.26
MDL Number MFCD00005411
Synonym Glyoxylic acid ethyl ester ethylene mercaptal