Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 1,2,3-thiadiazole-4-carboxylate, 97%

$99.14 - $375.19

Chemical Identifiers

Chemical Name or Material Ethyl 1,2,3-thiadiazole-4-carboxylate
CAS 3989-36-4
Molecular Formula C5H6N2O2S
Formula Weight 158.18
MDL Number MFCD00052100
Synonym 1,2,3-Thiadiazole-4-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1991203

  SDS
alfa aesar™
A1991203
1g Each for $99.14

AAA1991206

  SDS
alfa aesar™
A1991206
5g Each for $375.19
Description & Specifications

Specifications

Formula Weight 158.18
Chemical Name or Material Ethyl 1,2,3-thiadiazole-4-carboxylate
Assay 97%
Molecular Formula C5H6N2O2S
MDL Number MFCD00052100
CAS 3989-36-4
Synonym 1,2,3-Thiadiazole-4-carboxylic acid ethyl ester
Melting Point 83° to 84°C
Quantity 1g