missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 1,2,3,4-tetrahydroisoquinoline-3-carboxylate, 95%

$46.12 - $453.38

Chemical Identifiers

CAS 41234-43-9
Molecular Formula C12H15NO2
Molecular Weight (g/mol) 205.257
MDL Number MFCD04035517
InChI Key RGHMPTHWVVRXHW-UHFFFAOYSA-N
Synonym 1,2,3,4-tetrahydro-isoquinoline-3-carboxylic acid ethyl ester, 1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid ethyl ester, ethyl 1,2,3,4-tetrahydro-3-isoquinolinecarboxylate, 3-isoquinolinecarboxylic acid, 1,2,3,4-tetrahydro-, ethyl ester, 1,2,3,4-tetrahydroisoquinoline-3-carboxylic ethyl ester, 1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid ethyl ester, +/--1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid ethyl ester, 1,2,3,4-tetrahydro-3-isoquinoline carboxylic acid, ethyl ester, 3-isoquinolinecarboxylicacid, 1,2,3,4-tetrahydro-, ethyl ester
PubChem CID 652898
IUPAC Name ethyl 1,2,3,4-tetrahydroisoquinoline-3-carboxylate
SMILES CCOC(=O)C1CC2=CC=CC=C2CN1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH65722MD View Documents Alfa Aesar™
H65722MD
250mg Each for $46.12
 
AAH6572203 View Documents Alfa Aesar™
H6572203
1g Each for $121.51
 
AAH6572206 View Documents Alfa Aesar™
H6572206
5g Each for $453.38
 
Specifications

Chemical Identifiers

41234-43-9
205.257
RGHMPTHWVVRXHW-UHFFFAOYSA-N
652898
CCOC(=O)C1CC2=CC=CC=C2CN1
C12H15NO2
MFCD04035517
1,2,3,4-tetrahydro-isoquinoline-3-carboxylic acid ethyl ester, 1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid ethyl ester, ethyl 1,2,3,4-tetrahydro-3-isoquinolinecarboxylate, 3-isoquinolinecarboxylic acid, 1,2,3,4-tetrahydro-, ethyl ester, 1,2,3,4-tetrahydroisoquinoline-3-carboxylic ethyl ester, 1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid ethyl ester, +/--1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid ethyl ester, 1,2,3,4-tetrahydro-3-isoquinoline carboxylic acid, ethyl ester, 3-isoquinolinecarboxylicacid, 1,2,3,4-tetrahydro-, ethyl ester
ethyl 1,2,3,4-tetrahydroisoquinoline-3-carboxylate

Specifications

250mg
C12H15NO2
1,2,3,4-tetrahydro-isoquinoline-3-carboxylic acid ethyl ester, 1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid ethyl ester, ethyl 1,2,3,4-tetrahydro-3-isoquinolinecarboxylate, 3-isoquinolinecarboxylic acid, 1,2,3,4-tetrahydro-, ethyl ester, 1,2,3,4-tetrahydroisoquinoline-3-carboxylic ethyl ester, 1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid ethyl ester, +/--1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid ethyl ester, 1,2,3,4-tetrahydro-3-isoquinoline carboxylic acid, ethyl ester, 3-isoquinolinecarboxylicacid, 1,2,3,4-tetrahydro-, ethyl ester
CCOC(=O)C1CC2=CC=CC=C2CN1
205.257
205.26
95%
41234-43-9
MFCD04035517
RGHMPTHWVVRXHW-UHFFFAOYSA-N
ethyl 1,2,3,4-tetrahydroisoquinoline-3-carboxylate
652898
Solid
Ethyl 1,2,3,4-tetrahydroisoquinoline-3-carboxylate