missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 1,1,2,3,3,3-Hexafluoropropyl Ether, TCI America™

Manufacturer:  TCI America E102025G

 View more versions of this product

Catalog No. E102025G



Specifications

Specifications

Ethyl 1,1,2,3,3,3-Hexafluoropropyl Ether
Yellow
380-34-7
MFCD00236122
DMECHFLLAQSVAD-UHFFFAOYSA-N
1-ethoxy-1,1,2,3,3,3-hexafluoropropane
2775036
Clear Liquid at 20°C
65°C
25g
C5H6F6O
3271
CCOC(C(C(F)(F)F)F)(F)F
196.092
196.09
≥98.0% (GC)

Chemical Identifiers

380-34-7
196.092
DMECHFLLAQSVAD-UHFFFAOYSA-N
1-ethoxy-1,1,2,3,3,3-hexafluoropropane
C5H6F6O
MFCD00236122
2775036
CCOC(C(C(F)(F)F)F)(F)F
Safety and Handling

Safety and Handling

TSCA : No

Documents