Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 1,1,2,2-tetrafluoroethyl ether, 97%

Chemical Identifiers

Chemical Name or Material Ethyl 1,1,2,2-tetrafluoroethyl ether
CAS 512-51-6
Molecular Formula C4H6F4O
Linear Formula HCF2CF2OCH2CH3
Formula Weight 146.08
MDL Number MFCD00069162
UN Number UN3271
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2270306

 
alfa aesar™
B2270306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2270314

 
alfa aesar™
B2270314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 146.08
Chemical Name or Material Ethyl 1,1,2,2-tetrafluoroethyl ether
Assay 97%
Molecular Formula C4H6F4O
Linear Formula HCF2CF2OCH2CH3
MDL Number MFCD00069162
CAS 512-51-6
UN Number UN3271
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 56° to 58°C
Quantity 5g
Flash Point −15°C (−5°F)
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.