missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethoxytrimethylsilane, 97%

Manufacturer:  Alfa Aesar™ L1015122

 View more versions of this product

Catalog No. AAL1015122



Specifications

Specifications

Ethoxytrimethylsilane
C5H14OSi
UN1993
trimethylethoxysilane, silane, ethoxytrimethyl, silane, trimethylethoxy, ethyl trimethylsilyl ether, trimethylsilyl ethyl ether, ethoxy trimethyl silane, trimethyl ethoxy silane, silane,ethoxytrimethyl, acmc-1btam, ethanol, tms derivative
CCO[Si](C)(C)C
118.251
118.25
Mild
-83°C
−18°C (−0°F)
1825-62-3
MFCD00009069
1731950
RSIHJDGMBDPTIM-UHFFFAOYSA-N
ethoxy(trimethyl)silane
15772
97%
75°C to 76°C
100g

Chemical Identifiers

1825-62-3
118.251
RSIHJDGMBDPTIM-UHFFFAOYSA-N
15772
CCO[Si](C)(C)C
C5H14OSi
MFCD00009069
trimethylethoxysilane, silane, ethoxytrimethyl, silane, trimethylethoxy, ethyl trimethylsilyl ether, trimethylsilyl ethyl ether, ethoxy trimethyl silane, trimethyl ethoxy silane, silane,ethoxytrimethyl, acmc-1btam, ethanol, tms derivative
ethoxy(trimethyl)silane
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

RTECSNumber : VV4110000

TSCA : TSCA

Documents