missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethoxyquin, ≥90%, MP Biomedicals™
Manufacturer: Mp Biomedicals 0215796391
Specifications
Ethoxyquin | |
Brown | |
91-53-2 | |
C14H19NO | |
6-ethoxy-2,2,4-trimethyl-1,2-dihydroquinoline, antioxidant ec, ethoxyquin, ethoxyquine, niflex, santoflex a, santoflex aw, santoquin, santoquine, stop-scald | |
CCOC1=CC2=C(C=C1)NC(C=C2C)(C)C | |
217.312 | |
CHEBI:77323 | |
Viscous Liquid |
1.030g/mL | |
1kg | |
≥90% | |
MFCD00023883 | |
DECIPOUIJURFOJ-UHFFFAOYSA-N | |
6-ethoxy-2,2,4-trimethyl-1H-quinoline | |
3293 | |
217.3 |
Chemical Identifiers
91-53-2 | |
217.312 | |
DECIPOUIJURFOJ-UHFFFAOYSA-N | |
3293 | |
6-ethoxy-2,2,4-trimethyl-1H-quinoline |
C14H19NO | |
MFCD00023883 | |
6-ethoxy-2,2,4-trimethyl-1,2-dihydroquinoline, antioxidant ec, ethoxyquin, ethoxyquine, niflex, santoflex a, santoflex aw, santoquin, santoquine, stop-scald | |
CHEBI:77323 | |
CCOC1=CC2=C(C=C1)NC(C=C2C)(C)C |
Safety and Handling
EINECSNumber : 202-075-7
Recommended Storage : Store at Room Temperature (15 to 30°C). Store Desiccated. Store under nitrogen.