Learn More
Ethoxymethylenemalononitrile, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC117925000
Specifications
Ethoxymethylenemalononitrile | |
98% | |
C2H5OCH=C(CN)2 | |
03, I, 162 | |
OEICGMPRFOJHKO-UHFFFAOYSA-N | |
2-(ethoxymethylidene)propanedinitrile | |
67152 | |
Liquid | |
155g/mL | |
Glass bottle | |
63°C to 68°C |
123-06-8 | |
C6H6N2O | |
MFCD00001854 | |
2-cyano-3-ethoxyacrylonitrile, 2-ethoxymethylene malononitrile, 2-ethoxymethylidene propanedinitrile, ethoxymethylene malononitrile, ethoxymethylene propanedinitrile, ethoxymethylenemalononitrile, propanedinitrile, ethoxymethylene, usaf a-9230, usaf kf-10 | |
CCOC=C(C#N)C#N | |
122.127 | |
122.13 | |
≥97.5% (GC) | |
Authentic | |
Colorless to Yellow | |
500g |
Chemical Identifiers
123-06-8 | |
122.127 | |
OEICGMPRFOJHKO-UHFFFAOYSA-N | |
67152 | |
CCOC=C(C#N)C#N |
C6H6N2O | |
MFCD00001854 | |
2-cyano-3-ethoxyacrylonitrile, 2-ethoxymethylene malononitrile, 2-ethoxymethylidene propanedinitrile, ethoxymethylene malononitrile, ethoxymethylene propanedinitrile, ethoxymethylenemalononitrile, propanedinitrile, ethoxymethylene, usaf a-9230, usaf kf-10 | |
2-(ethoxymethylidene)propanedinitrile |
Safety and Handling
GHS H Statement:
Toxic in contact with skin.
Toxic if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger