Learn More
Ethoxymethylenemalononitrile, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 117921000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethoxymethylenemalononitrile | |
123-06-8 | |
155g/mL | |
Authentic | |
Liquid | |
C6H6N2O | |
MFCD00001854 | |
ethoxymethylenemalononitrile, ethoxymethylene malononitrile, propanedinitrile, ethoxymethylene, 2-ethoxymethylene malononitrile, 2-ethoxymethylidene propanedinitrile, 2-cyano-3-ethoxyacrylonitrile, ethoxymethylene propanedinitrile, usaf a-9230, usaf kf-10 | |
CCOC=C(C#N)C#N | |
122.127 | |
122.13 |
63°C to 68°C | |
Colorless to Yellow | |
100 g | |
Glass Bottle | |
98% | |
C2H5OCH=C(CN)2 | |
03, I, 162 | |
OEICGMPRFOJHKO-UHFFFAOYSA-N | |
2-(ethoxymethylidene)propanedinitrile | |
67152 | |
≥97.5% (GC) |
Chemical Identifiers
123-06-8 | |
122.127 | |
OEICGMPRFOJHKO-UHFFFAOYSA-N | |
67152 | |
CCOC=C(C#N)C#N |
C6H6N2O | |
MFCD00001854 | |
ethoxymethylenemalononitrile, ethoxymethylene malononitrile, propanedinitrile, ethoxymethylene, 2-ethoxymethylene malononitrile, 2-ethoxymethylidene propanedinitrile, 2-cyano-3-ethoxyacrylonitrile, ethoxymethylene propanedinitrile, usaf a-9230, usaf kf-10 | |
2-(ethoxymethylidene)propanedinitrile |
Safety and Handling
GHS H Statement:
Toxic in contact with skin.
Toxic if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The inform
Danger
RUO – Research Use Only