Please login to your online account to display your discounted pricing

Alfa Aesar™ (Ethoxycarbonylmethyl)triphenylphosphonium chloride, 97%

$126.08 - $417.15

Chemical Identifiers

Chemical Name or Material (Ethoxycarbonylmethyl)triphenylphosphonium chloride
CAS 17577-28-5
Molecular Formula CH3CH2O2CCH2P(C6H5)3Cl
Formula Weight 384.84
MDL Number MFCD00011746
Beilstein 3924139
Synonym (Carboethoxymethyl)triphenylphosphonium chloride; Ethyl (triphenylphosphonio)acetate chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1949414

  SDS
alfa aesar™
A1949414
25g Each for $126.08

AAA1949422

  SDS
alfa aesar™
A1949422
100g Each for $417.15
Description & Specifications

Specifications

Formula Weight 384.84
Chemical Name or Material (Ethoxycarbonylmethyl)triphenylphosphonium chloride
TSCA TSCA
Assay 97%
Molecular Formula CH3CH2O2CCH2P(C6H5)3Cl
MDL Number MFCD00011746
CAS 17577-28-5
Beilstein 3924139
Synonym (Carboethoxymethyl)triphenylphosphonium chloride; Ethyl (triphenylphosphonio)acetate chloride
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
Melting Point ≈145°C (dec.)
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C