missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethoxycarbonylmethyl(triphenyl)phosphonium Bromide, TCI America™

Manufacturer:  TCI America E0407250G

 View more versions of this product

Catalog No. E0407250G



Specifications

Specifications

Ethoxycarbonylmethyl(triphenyl)phosphonium Bromide
158°C
1530-45-6
MFCD00011835
VJVZPTPOYCJFNI-UHFFFAOYSA-M
(2-ethoxy-2-oxoethyl)-triphenylphosphanium;bromide
73731
Crystal-Powder at 20°C
Undesignated
250g
C22H22BrO2P
carbethoxymethyl triphenylphosphonium bromide, 2-ethoxy-2-oxoethyl triphenylphosphonium bromide, ethoxycarbonylmethyl triphenylphosphonium bromide, carbethoxymethylenetriphenylphosphonium bromide, ethoxycarbonylmethyl triphenyl phosphonium bromide, phosphonium, 2-ethoxy-2-oxoethyl triphenyl-, bromide, carbethoxymethyl-triphenyl-phosphonium bromide, 2-ethoxy-2-oxoethyl triphenylphosphanium bromide, carboethoxymethyl triphenylphosphonium bromide, phosphonium, ethoxycarbonylmethyl triphenyl-, bromide
CCOC(=O)C[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-]
429.294
429.29
≥97.0% (T)

Chemical Identifiers

1530-45-6
429.294
VJVZPTPOYCJFNI-UHFFFAOYSA-M
73731
CCOC(=O)C[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-]
C22H22BrO2P
MFCD00011835
carbethoxymethyl triphenylphosphonium bromide, 2-ethoxy-2-oxoethyl triphenylphosphonium bromide, ethoxycarbonylmethyl triphenylphosphonium bromide, carbethoxymethylenetriphenylphosphonium bromide, ethoxycarbonylmethyl triphenyl phosphonium bromide, phosphonium, 2-ethoxy-2-oxoethyl triphenyl-, bromide, carbethoxymethyl-triphenyl-phosphonium bromide, 2-ethoxy-2-oxoethyl triphenylphosphanium bromide, carboethoxymethyl triphenylphosphonium bromide, phosphonium, ethoxycarbonylmethyl triphenyl-, bromide
(2-ethoxy-2-oxoethyl)-triphenylphosphanium;bromide
Safety and Handling

Safety and Handling

RTECSNumber : TA2299500

TSCA : No

Documents