missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethoxycarbonyl isothiocyanate, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 187850050

 View more versions of this product

Catalog No. AC187850050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Ethoxycarbonyl isothiocyanate
1.1600g/mL
50°C
Glass bottle
SCNCOOCH2CH3
5g
04,223; 06,250; 07,146
ethoxycarbonyl isothiocyanate, ethyl isothiocyanatoformate, carbethoxy isothiocyanate, ethoxycarbonylisothiocyanate, ethyl n-carbothioylcarbamate, ethyl n-sulfanylidenemethylidene carbamate, ethyl isothiocyanatidocarbonate, carbon isothiocyanatidic acid, ethyl ester, o-ethyl carbonisothiocyanatidate
CCOC(=O)N=C=S
131.15
131.15
16182-04-0
56.0°C (18.0 mmHg)
94% min. (GC)
C4H5NO2S
MFCD00004814
03, III, 279
1.16
BDTDECDAHYOJRO-UHFFFAOYSA-N
ethyl N-(sulfanylidenemethylidene)carbamate
85320
95%

Chemical Identifiers

16182-04-0
131.15
BDTDECDAHYOJRO-UHFFFAOYSA-N
85320
CCOC(=O)N=C=S
C4H5NO2S
MFCD00004814
ethoxycarbonyl isothiocyanate, ethyl isothiocyanatoformate, carbethoxy isothiocyanate, ethoxycarbonylisothiocyanate, ethyl n-carbothioylcarbamate, ethyl n-sulfanylidenemethylidene carbamate, ethyl isothiocyanatidocarbonate, carbon isothiocyanatidic acid, ethyl ester, o-ethyl carbonisothiocyanatidate
ethyl N-(sulfanylidenemethylidene)carbamate
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor

GHS Signal Word: Danger

EINECSNumber : 240-318-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only