missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethoxycarbonyl isocyanate, 90%, tech., Thermo Scientific™
Supplier: Thermo Scientific Chemicals 314310250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Ethoxycarbonyl isocyanate | |
1.1100g/mL | |
35°C | |
88% min. (GC) | |
C4H5NO3 | |
1.4060 to 1.4090 | |
1.11 | |
VMVZGGPZNHFGKS-UHFFFAOYSA-N | |
ethyl N-(oxomethylidene)carbamate | |
300166 | |
90% |
19617-43-7 | |
25.0°C (10.0 mmHg) | |
Authentic | |
Glass bottle | |
C2H5O2CNCO | |
25g | |
ethoxycarbonyl isocyanate, ethyl isocyanatoformate, ethyl carbonisocyanatidate, ethyl n-carbonylcarbamate, ethoxycarbonylisocyanate, carbonisocyanatidic acid, ethyl ester, ethyl isocyanatooate, ethylisocyanatoformate, acmc-20al2o, carbonisocyanatidicacid,ethylester | |
CCOC(=O)N=C=O | |
115.09 | |
115.09 |
Safety and Handling
GHS Signal Word: Warning
RUO â Research Use Only