Learn More
Ethoxyacetic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2419422
Description
Ethoxyacetic acid is used to evaluate toxicity in embryonic stem cell test in mice. It finds application as a biomarker of exposure to alkyl-substituted ethylene glycols and other aliphatic ethers. It is used as a precursor to prepare ethoxy-acetic acid ethoxymethyl ester.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthoxyacetic acid is used to evaluate toxicity in embryonic stem cell test in mice. It finds application as a biomarker of exposure to alkyl-substituted ethylene glycols and other aliphatic ethers. It is used as a precursor to prepare ethoxy-acetic acid ethoxymethyl ester.
Solubility
Miscible with water.
Notes
Incompatible with bases, oxidizing agents and reducing agents.
Specifications
Ethoxyacetic acid | |
1.102 | |
97°C (206°F) | |
C4H8O3 | |
MFCD00004310 | |
UN3265 | |
ethoxyacetic acid, acetic acid, ethoxy, ethoxy acetic acid, acetic acid, 2-ethoxy, ethoxy-acetic acid, unii-467vw095bx, o-ethylglycolic acid, ccris 7194, 2-ethoxy-acetic acid, dsstox_cid_11294 | |
YZGQDNOIGFBYKF-UHFFFAOYSA-N | |
2-ethoxyacetic acid | |
12301 | |
98% |
627-03-2 | |
207°C | |
Pungent | |
1.419 | |
100 g | |
1743037 | |
Miscible with water. | |
CCOCC(=O)O | |
104.105 | |
104.11 |
Chemical Identifiers
627-03-2 | |
104.105 | |
YZGQDNOIGFBYKF-UHFFFAOYSA-N | |
12301 | |
CCOCC(=O)O |
C4H8O3 | |
MFCD00004310 | |
ethoxyacetic acid, acetic acid, ethoxy, ethoxy acetic acid, acetic acid, 2-ethoxy, ethoxy-acetic acid, unii-467vw095bx, o-ethylglycolic acid, ccris 7194, 2-ethoxy-acetic acid, dsstox_cid_11294 | |
2-ethoxyacetic acid |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 210-978-2
RTECSNumber : AH3600000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only