Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethidium Bromide, Molecular Biology Grade, 98%


Manufacturer: alfa aesar™  J6727006

 View more versions of this product

Catalog No. AAJ6727006

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Assay 98%
Format Powder
Product Type Ethidium Bromide
Quantity 5G
Color Red to purple
Purity Molecular Biology Grade
Assay 98%
CAS 1239-45-8
Molecular Formula C21H20BrN3
Formula Weight 394.32
Synonym 3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide; Homidium bromide
Odor Weak
Melting Point ca 261° dec.