missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethidium bromide, 98% (dry wt.)

$34.01 - $133.15

Chemical Identifiers

CAS 1239-45-8
Molecular Formula C21H20BrN3
Formula Weight 394.32
MDL Number MFCD00011724
UN Number UN2811
Beilstein 3642536
Merck Index 14,4731
Synonym 3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide; Homidium bromide
Chemical Name or Material Ethidium bromide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0748206 SDS Alfa Aesar™
L0748206*
5g Each for $133.15
AAL0748203 SDS Alfa Aesar™
L0748203
1g Each for $34.01
Specifications Description & Specifications

Chemical Identifiers

1239-45-8
394.32
UN2811
14,4731
Ethidium bromide
C21H20BrN3
MFCD00011724
3642536
3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide; Homidium bromide

Specifications

1239-45-8
394.32
UN2811
14,4731
≈261°C
5g
98%, (dry wt.)
C21H20BrN3
MFCD00011724
3642536
3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide; Homidium bromide
Weak
Ethidium bromide

Safety and Handling

GHS H Statement
H330-H341-H302
Fatal if inhaled.
Suspected of causing genetic defects.
Harmful if swallowed.

GHS P Statement
P260-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : SF7950000