Please login to your online account to display your discounted pricing

Ether USP, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Ether
CAS 60-29-7
Purity Grade BP/EP/FCC/JP/NF/USP
Molecular Formula (C2H5)2O
Formula Weight 74.12
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK0812212

 
avantor performance
081216
Al Bottle 2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK0812500

 
avantor performance
081210
Al Bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 74.12
Purity Grade BP/EP/FCC/JP/NF/USP
Quantity 2.5kg
Chemical Name or Material Ether
Molecular Formula (C2H5)2O
CAS 60-29-7
Packaging Al Bottle