Please login to your online account to display your discounted pricing

Ether, Anhydrous, HPLC Grade, J.T.Baker™

High purity solvents for use in analytical and preparative separations

$379.15 - $1,287.26

Chemical Identifiers

Chemical Name or Material Ether
CAS 60-29-7
Name Note Anhydrous
Purity Grade HPLC Grade
Purity Grade Notes Meets USP requirements
Molecular Formula (C2H5)2O
Formula Weight 74.12
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002818

 
avantor performance
923703
Narrow Mouth Amber Glass Bottle 4L Each for $1,287.26

02002819

 
avantor performance
923733
Narrow Mouth PVC Coated Amber Glass Bottle 4L Each for $379.15
Description & Specifications

Specifications

Formula Weight 74.12
Purity Grade Notes Meets USP requirements
Flash Point −45.00°C (113°F)
Chemical Name or Material Ether
Name Note Anhydrous
Molecular Formula (C2H5)2O
Specific Gravity 1L = 0.71kg
Purity Grade HPLC Grade
CAS 60-29-7
Packaging Narrow Mouth Amber Glass Bottle
Quantity 4L

    For use in liquid chromatography (HPLC & UHPLC) & spectrophotometry, bulk pharmaceutical chemical