missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethanolamine (Laboratory), Fisher Chemical

$294.15 - $786.62

Chemical Identifiers

CAS 141-43-5
Molecular Formula C2H7NO
Linear Formula HOCH2CH2NH2
Formula Weight 61.08
MDL Number MFCD00008183
Synonym 2-Aminoethanol
Chemical Name or Material Ethanolamine
Grade Laboratory Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
M251-1 SDS Fisher Scientific Company
M2511
Glass Bottle 1L Each for $294.15
Case of 6 Each for $1,764.90
M251-4 SDS Accessories Fisher Scientific Company
M2514
Glass Bottle 4L Each for $786.62
Case of 4 Each for $3,146.48
Specifications

Description

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Chemical Identifiers

141-43-5
HOCH2CH2NH2
MFCD00008183
Ethanolamine
C2H7NO
61.08
2-Aminoethanol
Laboratory Grade

Specifications

Vapor Pressure: 0.48mmHg at 20°C
Liquid
C2H7NO
61.08
2491
Glass Bottle
2.1g/mol
170°C
12
1L
Ethanolamine
141-43-5
Pass Test
HOCH2CH2NH2
MFCD00008183
2-Aminoethanol
0.48mmHg at 20°C
1.012g/cm³
10°C
Colorless
92°C
Laboratory Grade

Safety and Handling

DANGER!

Emergency Overview
Combustible liquid. Harmful by inhalation, in contact with skin and if swallowed. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:3
Flammability:2
Instability:1

DOTInformation : DOT Class 8, : Corrosive