Please login to your online account to display your discounted pricing

Ethanolamine (Laboratory), Fisher Chemical


Quantity 4L
Packaging Glass Bottle

Manufacturer: fisher scientific company  M2514

 View more versions of this product

Catalog No. M251-4

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Laboratory Grade
Formula Weight 61.08
Boiling Point 170°C
Melting Point 10°C
Quantity 4L
UN Number 2491
Flash Point 92°C
DOT Information DOT Class 8, : Corrosive
Physical Form Liquid
Vapor Pressure 0.48mmHg at 20°C
Density 1.012g/cm³
pH 12
Color Colorless
Health Hazard 3 Emergency Overview
Combustible liquid. Harmful by inhalation, in contact with skin and if swallowed. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:3
Flammability:2
Instability:1
Chemical Name or Material Ethanolamine
Identification Pass Test
Molecular Formula C2H7NO
MDL Number MFCD00008183
Additional Information Vapor Pressure: 0.48mmHg at 20°C
CAS 141-43-5
ChemAlert Storage Symbol White
Synonym 2-Aminoethanol
Packaging Glass Bottle
Health Hazard 2 DANGER!
Linear Formula HOCH2CH2NH2

Specifications

Chemical Name or Material Ethanolamine
CAS 141-43-5
Purity Grade Laboratory Grade
Molecular Formula C2H7NO
Linear Formula HOCH2CH2NH2
Formula Weight 61.08
MDL Number MFCD00008183
Synonym 2-Aminoethanol
Accessories

Accessories for Catalog Number: M2514

Item Mfr. No. Item Description Price Per Unit Quantity  
Accessories
 

Emergency Overview
Combustible liquid. Harmful by inhalation, in contact with skin and if swallowed. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:3
Flammability:2
Instability:1

DANGER!

SDS & Safety Documents