Please login to your online account to display your discounted pricing

Ethanolamine, 99%, AcroSeal™, ACROS Organics™


Quantity 800mL
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  AC427258000

 View more versions of this product

Catalog No. AC427258000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 61.08
Physical Form Liquid
Quantity 800mL
Color Clear
Packaging Glass Bottle
Density 1.0120g/mL
Merck Index 15, 3782
Refractive Index 1.4530 to 1.4550 (20°C, 589nm)
Linear Formula H2NCH2CH2OH
Synonym 2-Aminoethanol
Infrared Spectrum Authentic
CAS 141-43-5
MDL Number MFCD00008183
Assay Percent Range 99%
Molecular Formula C2H7NO
Chemical Name or Material Ethanolamine
Health Hazard 1 Danger
Water 0.5% max.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
May cause respiratory irritation.

Specifications

Chemical Name or Material Ethanolamine
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C2H7NO
Linear Formula H2NCH2CH2OH
Formula Weight 61.08
MDL Number MFCD00008183
Synonym 2-Aminoethanol
Accessories

Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
May cause respiratory irritation.

SDS & Safety Documents