Please login to your online account to display your discounted pricing

Ethanolamine, 99%, AcroSeal™, ACROS Organics™


Quantity 100mL
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  AC427251000

 View more versions of this product

Catalog No. AC427251000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥98.5%
Formula Weight 61.08
Quantity 100mL
Color Clear
Merck Index 15, 3782
Packaging Glass Bottle
Density 1.0120g/mL
Linear Formula H2NCH2CH2OH
Synonym 2-Aminoethanol
Water 0.5% max.
Chemical Name or Material Ethanolamine
Assay Percent Range 99%
Infrared Spectrum Authentic
MDL Number MFCD00008183
CAS 141-43-5
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C2H7NO
Health Hazard 1 Danger
Refractive Index 1.4530 to 1.4550 (20°C, 589nm)

Specifications

Chemical Name or Material Ethanolamine
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C2H7NO
Linear Formula H2NCH2CH2OH
Formula Weight 61.08
MDL Number MFCD00008183
Synonym 2-Aminoethanol
Accessories

Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
May cause respiratory irritation.

SDS & Safety Documents