Please login to your online account to display your discounted pricing

Ethanol, SIS Formula A-1, 200 Proof, PHARMCO-AAPER™

Applications include Industrial and manufacturing processes

$5,499.89 - $5,499.89

Chemical Identifiers

CAS 64-17-5
CAS 67-63-0
CAS 67-56-1
CAS 108-10-1
Physical Form Liquid
Molecular Formula C2H6O
Formula Weight 46.07
Synonym Denatured Ethanol
View More Specs

 Alert

Minimum Order Quantity: 5

Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

16-101-143

 
labconco corp
LCN3780102
Metal pail 5 gal. Each for $5,499.89
Description & Specifications

Specifications

Physical Form Liquid
Formula Weight 46.07
Color Clear colorless
Quantity 5 gal.
Molecular Formula C2H6O
CAS 64-17-5
Synonym Denatured Ethanol
Decomposition Information Carbon oxides produce hazardous products under combustion conditions
Stability Stable under recommended storage conditions.
Packaging Metal pail
CAS 108-10-1
Health Hazard 1 GHS Signal Word: Danger
CAS 67-63-0
CAS 67-56-1
Physical State Liquid
Safety and Handling

GHS Signal Word: Danger