Please login to your online account to display your discounted pricing

Ethanol, denatured, special, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Ethanol
CAS 64-17-5
CAS 67-56-1
CAS 141-78-6
Physical Form Liquid
Molecular Formula C2H6O
Formula Weight 46.08
MDL Number MFCD00003568
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41603K2

  SDS
alfa aesar
41603K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear colorless
Physical Form Liquid
Formula Weight 46.08
Quantity 1L
DOT Information DOT Class: 3, Packaging Group: II
EINECS Number 200-578-6
Density 0.7893g/mL
Concentration or Composition (by Analyte or Components) Ethyl alcohol: 92%; Methanol: 4%; Ethyl acetate: 4%
CAS 67-56-1
CAS 64-17-5
CAS 141-78-6
Chemical Name or Material Ethanol
Odor Alcohol-like
MDL Number MFCD00003568
Molecular Formula C2H6O
Appearance Colorless liquid
Health Hazard 1 H225-H371-H333: Highly flammable liquid and vapour. May cause damage to organs. May be harmful if inhaled.
Safety and Handling

H225-H371-H333: Highly flammable liquid and vapour. May cause damage to organs. May be harmful if inhaled.