Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethanol-d6, 99.5%(Isotopic)

Chemical Identifiers

Chemical Name or Material Ethanol-d6
CAS 1516-08-1
Name Note Isotopic
Linear Formula CD3CD2OD
Formula Weight 52.11g/mol
MDL Number MFCD00051020
UN Number UN1170
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3650203

 
alfa aesar™
36502-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3650206

 
alfa aesar™
36502-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Odor Alcohol-like
Quantity 1g
Sensitivity Hygroscopic
Density 0.91g/mL
Boiling Point 79°C
Quantity 1g
Color Colorless
Chemical Name or Material Ethanol-d6
Name Note Isotopic
CAS 1516-08-1
Physical Form Liquid
Assay 99.5%
Linear Formula CD3CD2OD
Formula Weight 52.11g/mol
MDL Number MFCD00051020
UN Number UN1170
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHANOL SOLUTIONS
Safety and Handling

Danger

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225
Highly flammable liquid and vapor.