Please login to your online account to display your discounted pricing

Ethanol-Benzene Mixture, 1:2, LabChem™

$117.54 - $117.54

Chemical Identifiers

Chemical Name or Material Ethanol-Benzene Mixture
CAS 71-43-2
CAS Max % 66.67
CAS 64-17-5
CAS Max % 30
CAS 67-63-0
CAS Max % 1.665
CAS 67-56-1
CAS Max % 1.665
Name Note 1:2
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC141004

 
lab chem
LC141004
4L Each for $117.54
Description & Specifications

Specifications

Formula Weight 78.11
Chemical Name or Material Ethanol-Benzene Mixture
Identification Passes Test
CAS Max % 1.665
CAS 67-56-1
CAS Max % 1.665
Packaging Amber Glass
Storage Information Room Temperature
Name Note 1:2
Decomposition Information Carbon monoxide; Carbon dioxide; May release flammable gases
Physical Form Liquid
Color Clear, colorless
Quantity 4L
Linear Formula C6H6
CAS 71-43-2
UN Number UN1993
Health Hazard 1 Danger
CAS Max % 66.67
Health Hazard 2 GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes skin and serious eye irritation.
May cause genetic defects.
May cause cancer.
Suspected of damaging fertility or the unborn child.
Causes damage to organs (central nervous system, optic nerve).
Causes damage to organs (liver, kidneys) through prolonged or repeated exposure.
Toxic to aquatic life.
CAS 64-17-5
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
If skin irritation occurs: Get medical advice/attention.
Wash contaminated clothing before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
In case of fire: Use alcohol resistant foam, carbon dioxide, extinguishing powder to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
CAS Max % 30
CAS 67-63-0
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
If skin irritation occurs: Get medical advice/attention.
Wash contaminated clothing before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
In case of fire: Use alcohol resistant foam, carbon dioxide, extinguishing powder to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes skin and serious eye irritation.
May cause genetic defects.
May cause cancer.
Suspected of damaging fertility or the unborn child.
Causes damage to organs (central nervous system, optic nerve).
Causes damage to organs (liver, kidneys) through prolonged or repeated exposure.
Toxic to aquatic life.