Please login to your online account to display your discounted pricing

Ethanol, anhydrous, denatured, Spectrophotometric Grade, 90%, 5% methanol, 5% isopropanol, Alfa Aesar™

$421.11 - $421.11

Chemical Identifiers

Chemical Name or Material Ethanol
CAS 64-17-5
Purity Grade Spectrophotometric Grade
Physical Form Liquid
Assay Percent Range 90%
Molecular Formula C2H6O
Formula Weight 46.08g/mol
MDL Number MFCD00003568
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA22931M6

 
alfa aesar
22931-M6
4 x 4L Each for $421.11
Description & Specifications

Specifications

Density 0.789
Quantity 4 x 4L
Chemical Name or Material Ethanol
CAS 64-17-5
Purity Grade Spectrophotometric Grade
Physical Form Liquid
Assay Percent Range 90%
Molecular Formula C2H6O
Formula Weight 46.08g/mol
MDL Number MFCD00003568
EINECS Number 200-578-6