Please login to your online account to display your discounted pricing

Ethanol, anhydrous, denatured, HPLC Grade, 90%, 5% methanol, 5% isopropanol, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Ethanol
CAS 64-17-5
Purity Grade HPLC Grade
Physical Form Liquid
Assay Percent Range 90%
Molecular Formula C2H6O
Formula Weight 46.08
MDL Number MFCD00003568
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA22930M6

  SDS
alfa aesar
22930M6
4 x 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point −90°C
Boiling Point 78°C
Purity Grade HPLC Grade
Formula Weight 46.08
Quantity 4 x 4L
Color Colorless
DOT Information DOT Class: 3; Packing Group: II
Refractive Index 1.3610
UN Number 1987
Chemical Name or Material Ethanol
Physical Form Liquid
CAS 64-17-5
Density 0.789g/cm3 at 20°C
EINECS Number 200-578-6
Assay Percent Range 90%
Appearance Colorless liquid
Health Hazard 1 H225-H371-H333: Highly flammable liquid and vapour. May cause damage to organs. May be harmful if inhaled.
Molecular Formula C2H6O
Vapor Pressure 57 hPa (43mm Hg) at 20°C
Odor Alcohol-like
MDL Number MFCD00003568