Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethanol, anhydrous, denatured, 85.8%, 13.3% methanol, 0.9% MIBK

Chemical Identifiers

Chemical Name or Material Ethanol
CAS 64-17-5
Name Note Anhydrous, denatured
Molecular Formula C2H6O
Linear Formula CH3CH2OH
Formula Weight 46.08g/mol
MDL Number MFCD00003568
UN Number UN1993
Beilstein 1718733
Merck Index 14,3760
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA36642AP

 
alfa aesar™
36642-AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Concentration 13.3% methanol, 0.9% MIBK
Density 0.811g/mL
Odor Like alcohol
Refractive Index 1.3610
Flash Point 11° (52°F)
Boiling Point 80°C
Color Colorless
Melting Point −100°C
Quantity 500mL
Sensitivity Hygroscopic
Chemical Name or Material Ethanol
Name Note Anhydrous, denatured
CAS 64-17-5
Physical Form Liquid
Assay 85.8%
Assay Percent Range 85.8%
Molecular Formula C2H6O
Linear Formula CH3CH2OH
Formula Weight 46.08g/mol
MDL Number MFCD00003568
UN Number UN1993
Beilstein 1718733
Merck Index 14,3760
Synonym Ethyl alcohol
Solubility Information Miscible in water and most organic solvents
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H370
Highly flammable liquid and vapor.
Causes damage to organs.
Health Hazard 3 GHS P Statement
P210-P260-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
RTECS Number KQ6300000
TSCA TSCA

  • As a gasoline additive/substitute; octane booster
  • Very mobile liquid solvent
  • Used in lotions, tonics and perfumes

Safety and Handling

Danger

GHS P Statement
P210-P260-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.

GHS H Statement
H225-H370
Highly flammable liquid and vapor.
Causes damage to organs.