Please login to your online account to display your discounted pricing

Ethanethiol, 97%, Alfa Aesar™

$37.22 - $37.22

Chemical Identifiers

Chemical Name or Material Ethanethiol
CAS 75-08-1
Molecular Formula C2H6S
Linear Formula CH3CH2SH
Formula Weight 62.13
MDL Number MFCD00004887
UN Number UN2363
Beilstein 773638
Merck Index 14,3726
Synonym Ethyl mercaptan
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA22585AE

  SDS
alfa aesar
22585AE
Each for $37.22
Description & Specifications

Specifications

Formula Weight 62.13
Quantity 100mL
Chemical Name or Material Ethanethiol
TSCA TSCA
Physical Form Liquid
Assay 97%
Molecular Formula C2H6S
Linear Formula CH3CH2SH
MDL Number MFCD00004887
CAS 75-08-1
UN Number UN2363
Beilstein 773638
Merck Index 14,3726
Synonym Ethyl mercaptan
Solubility Information Soluble in alcohol, ether, water
Health Hazard 1 Danger
Refractive Index 1.4310
Health Hazard 2 GHS H Statement
H225-H332;
Highly flammable liquid and vapor.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P210-P261-P280-P303+P361+P353-P403+P235-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 0.84
Boiling Point 35°C
Melting Point −148°C
Color Colorless
Odor Unpleasant
Flash Point −48°C (−55°F)
DOT Information Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: ETHYL MERCAPTAN
RTECS Number KI9625000
Safety and Handling

Danger

GHS P Statement
P210-P261-P280-P303+P361+P353-P403+P235-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H332;
Highly flammable liquid and vapor.
Harmful if inhaled.