missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethanethiol, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 022585AP

 View more versions of this product

Catalog No. AA22585AP


Description

Description

Ethanethiol acts as a stabilizer for adhesives and an odorant for natural gas. It is used in the odorless liquefied petroleum gas (LPG) in order to find out the gas leaks. It is used as key starting material for the preparation of ethylsulfonic acid, diethyldisulfide and sodium ethylmercaptide. Further, it is also used in the underground mining industry as stench gas. In organic synthesis, it gives powerful nucleophile set- in the presence of sodium hydroxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethanethiol
-148°C
0.84 g/mL
-48°C (-55°F)
C2H6S
CH3CH2SH
500 mL
773638
ethyl mercaptan, mercaptoethane, thioethanol, ethyl hydrosulfide, ethyl sulfhydrate, ethyl thioalcohol, thioethyl alcohol, 1-mercaptoethane, aethanethiol, aethylmercaptan
DNJIEGIFACGWOD-UHFFFAOYSA-N
ethanethiol
6343
97%
75-08-1
Colorless
35°C
Unpleasant
1.431
MFCD00004887
UN2363
14,3726
Soluble in alcohol,ether,waterMiscible with alcohol,petroleum naphtha,acetone,dilute alkali and ether; Slightly miscible with water.
CCS
62.13
62.13
Liquid

Chemical Identifiers

75-08-1
62.13
DNJIEGIFACGWOD-UHFFFAOYSA-N
6343
CCS
C2H6S
MFCD00004887
ethyl mercaptan, mercaptoethane, thioethanol, ethyl hydrosulfide, ethyl sulfhydrate, ethyl thioalcohol, thioethyl alcohol, 1-mercaptoethane, aethanethiol, aethylmercaptan
ethanethiol
Safety and Handling

Safety and Handling

GHS H Statement
H225-H332
Highly flammable liquid and vapor.
Harmful if inhaled.

P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P272-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P333+P313-P363-P370+P378q-P501c

H225-H302+H332-H317

DOTInformation : Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: ETHYL MERCAPTAN

EINECSNumber : 200-837-3

RTECSNumber : KI9625000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only