missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethanesulfonyl chloride, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 215820050

 View more versions of this product

Catalog No. AC215820050



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethanesulfonyl chloride
594-44-5
1.3500g/mL
83°C
98% min. (GC)
C2H5ClO2S
C2H5SO2Cl
5 mL
1.35
Solubility in water: decomposes in water. Other solubilities: soluble in acetone and ethyl ether
CCS(Cl)(=O)=O
128.57
1.44 mm2/s (20°C)
98+%
98+%
Brown to Pink-Purple or Colorless to Yellow
177.0°C
Authentic
Glass bottle
1.4520 to 1.4540
MFCD00007460
04, 6
ethylsulfonyl chloride, ethylsulfochloride, 1-ethanesulfonyl chloride, ethanesulphonyl chloride, ethanesulfonylchloride, unii-b56zoc898a, ethylsulfonylchloride, ethansulfonyl chloride, ethansulfonylchlorid, ethanesulfonylchoride
FRYHCSODNHYDPU-UHFFFAOYSA-N
ethanesulfonyl chloride
11667
128.58
Liquid

Chemical Identifiers

594-44-5
128.57
FRYHCSODNHYDPU-UHFFFAOYSA-N
11667
CCS(Cl)(=O)=O
C2H5ClO2S
MFCD00007460
ethylsulfonyl chloride, ethylsulfochloride, 1-ethanesulfonyl chloride, ethanesulphonyl chloride, ethanesulfonylchloride, unii-b56zoc898a, ethylsulfonylchloride, ethansulfonyl chloride, ethansulfonylchlorid, ethanesulfonylchoride
ethanesulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if inhaled.
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-842-5

RTECSNumber : KI8050000

TSCA : TSCA

Recommended Storage : this product darkens during storage

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only