Please login to your online account to display your discounted pricing

Ethanesulfonic acid, 95%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC366040050

 View more versions of this product

Catalog No. AC366040050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Liquid
Formula Weight 110.13
Quantity 5g
Color Clear yellow to brown
Beilstein 04, 5
Health Hazard 3 GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Density 1.3430g/mL
Assay Percent Range 95%
Packaging Glass bottle
Chemical Name or Material Ethanesulfonic acid
Molecular Formula C2H6O3S
Refractive Index 1.4325 to 1.4355 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 1 Danger
Name Note 95%
Linear Formula C2H5SO3H
MDL Number MFCD00007529
CAS 594-45-6
Health Hazard 2 GHS P Statement
Causes severe skin burns and eye damage.

Specifications

Chemical Name or Material Ethanesulfonic acid
Name Note 95%
CAS 594-45-6
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C2H6O3S
Linear Formula C2H5SO3H
Formula Weight 110.13
MDL Number MFCD00007529
Accessories

Danger

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS P Statement
Causes severe skin burns and eye damage.

SDS & Safety Documents