missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethambutol dihydrochloride, Thermo Scientific Chemicals

An antibacterial agent

Supplier:  Thermo Scientific Chemicals J6069506

 View more versions of this product

Catalog No. AAJ6069506



Description

Description

Ethambutol dihydrochloride it acts as a tuberculostatic antibacterial agent. It is used to treat tuberculosis in combination with other antibiotics to reduce the development of drug resistance. It has been used to study multidrug-resistant tuberculosis (MDR-TB) in highest-incidence-rate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Slightly soluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications

Specifications

Ethambutol dihydrochloride
White
MFCD00216025
14,3720
Slightly soluble in water
CCC(CO)NCCNC(CC)CO.Cl.Cl
277.23
277.23
1070-11-7
C10H26Cl2N2O2
5 g
ethambutol 2hcl, 2r-2-2-2r-1-hydroxybutan-2-yl amino ethyl amino butan-1-ol dihydrochloride, ethambutol related compound b usp, prestwick_208, aronis24117, 2,2'-1,2-ethanediyldiimino bis-1-butanol dihydrochloride, unii-jng307dj5x component, 2,2'-1,2-ethanediyldi imino di 1-butanol dihydrochloride, 2r-2-2-2r-1-hydroxybutan-2-yl amino ethylamino butan-1-ol dihydrochloride, 2r,2'r-2,2'-ethane-1,2-diylbis azanediyl bis butan-1-ol dihydrochloride
AUAHHJJRFHRVPV-HSTMFJOWSA-N
(2R)-2-[2-[[(2R)-1-hydroxybutan-2-yl]amino]ethylamino]butan-1-ol;dihydrochloride
6419929
Powder

Chemical Identifiers

1070-11-7
277.23
AUAHHJJRFHRVPV-HSTMFJOWSA-N
6419929
CCC(CO)NCCNC(CC)CO.Cl.Cl
C10H26Cl2N2O2
MFCD00216025
ethambutol 2hcl, 2r-2-2-2r-1-hydroxybutan-2-yl amino ethyl amino butan-1-ol dihydrochloride, ethambutol related compound b usp, prestwick_208, aronis24117, 2,2'-1,2-ethanediyldiimino bis-1-butanol dihydrochloride, unii-jng307dj5x component, 2,2'-1,2-ethanediyldi imino di 1-butanol dihydrochloride, 2r-2-2-2r-1-hydroxybutan-2-yl amino ethylamino butan-1-ol dihydrochloride, 2r,2'r-2,2'-ethane-1,2-diylbis azanediyl bis butan-1-ol dihydrochloride
(2R)-2-[2-[[(2R)-1-hydroxybutan-2-yl]amino]ethylamino]butan-1-ol;dihydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H360
May damage fertility or the unborn child.

P201-P202-P281-P308+P313-P501c

H360

EINECSNumber : 213-970-7

RTECSNumber : EL3854000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only