missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethambutol, 98%, Alfa Aesar™

Used in conjunction with at least one other antituberculous drug in the treatment of pulmonary tuberculosis.

Manufacturer:  Thermo Fisher Scientific J6614522

 View more versions of this product

Catalog No. AAJ6614522



Description

Description

Solubility
Soluble in water. Also soluble in chloroform and methylene chloride

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Specifications

Specifications

Ethambutol
C10H24N2O2
Glass bottle
CCC(CO)NCCNC(CC)CO
204.314
CHEBI:4877
Powder
345°C
100g
74-55-5
MFCD00242828
AEUTYOVWOVBAKS-UWVGGRQHSA-N
(2S)-2-[2-[[(2S)-1-hydroxybutan-2-yl]amino]ethylamino]butan-1-ol
14052
204.31
98%
199°C to 204°C
>110°C

Chemical Identifiers

74-55-5
204.314
AEUTYOVWOVBAKS-UWVGGRQHSA-N
CHEBI:4877
CCC(CO)NCCNC(CC)CO
C10H24N2O2
MFCD00242828
14052
(2S)-2-[2-[[(2S)-1-hydroxybutan-2-yl]amino]ethylamino]butan-1-ol
Safety and Handling

Safety and Handling

TSCA : No

Documents