Please login to your online account to display your discounted pricing

Etazolate hydrochloride, Tocris


PDE4 inhibitor

Quantity 10mg

Manufacturer: r & d systems  043810

 View more versions of this product

Catalog No. 043810

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 325.8
Assay Percent Range >98%
Chemical Name or Material Etazolate hydrochloride
CAS 35838-58-5
Synonym 1-Ethyl-4-[(1-methylethylidene)hydrazino]-1H-pyrazolo-[3, 4-b]-pyridine-5-carboxylic acid, ethyl ester hydrochloride
Molecular Formula C14H19N5O2·HCl
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material Etazolate hydrochloride
Assay Percent Range >98%
Molecular Formula C14H19N5O2·HCl
Formula Weight 325.8
Synonym 1-Ethyl-4-[(1-methylethylidene)hydrazino]-1H-pyrazolo-[3, 4-b]-pyridine-5-carboxylic acid, ethyl ester hydrochloride
Accessories

Store at Room Temperature