missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Etamsylate, TCI America™

Manufacturer:  TCI America E11455G

 View more versions of this product

Catalog No. E11455G



Specifications

Specifications

Etamsylate
130°C
2624-44-4
MFCD00867499
HBGOLJKPSFNJSD-UHFFFAOYSA-N
2,5-dihydroxybenzenesulfonic acid;N-ethylethanamine
17506
Crystal-Powder at 20°C
White
5g
C10H17NO5S
Diethylamine 2,5-Dihydroxybenzenesulfonate
CCNCC.C1=CC(=C(C=C1O)S(=O)(=O)O)O
263.308
263.31
≥98.0% (HPLC,T)

Chemical Identifiers

2624-44-4
263.308
HBGOLJKPSFNJSD-UHFFFAOYSA-N
17506
CCNCC.C1=CC(=C(C=C1O)S(=O)(=O)O)O
C10H17NO5S
MFCD00867499
Diethylamine 2,5-Dihydroxybenzenesulfonate
2,5-dihydroxybenzenesulfonic acid;N-ethylethanamine
Safety and Handling

Safety and Handling

RTECSNumber : DB6145000

TSCA : Yes

Documents