Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Estrogen Sulfotransferase (Anti-Rat) Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry and immunofluorescence experiments.

$512.55 - $1,485.20

Specifications

Antigen Estrogen Sulfotransferase (Anti-Rat) Ab-1
Clone EST.1
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunofluorescence
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS233P

 
thermo scientific™ lab vision™
MS233P
Purified Ab with BSA and Azide 1mL Each for $1,382.61

MS233P0

 
thermo scientific™ lab vision™
MS233P0
Purified Ab with BSA and Azide 100µL Each for $512.55

MS233P1

 
thermo scientific™ lab vision™
MS233P1
Purified Ab with BSA and Azide 500µL Each for $1,033.67

MS233P1ABX

 
thermo scientific™ lab vision™
MS233P1ABX
Ab without BSA/Azide (Frozen) 100µL Each for $1,171.45

MS233PABX

 
thermo scientific™ lab vision™
MS233PABX
Ab wWithout BSA/Azide (Frozen) 200µL Each for $1,485.20

MS233R7

 
thermo scientific™ lab vision™
MS233R7
Ready-to-use for IHC (UVLP) 7mL Each for $1,083.06
Description & Specifications

Specifications

Antigen Estrogen Sulfotransferase (Anti-Rat) Ab-1
Clone EST.1
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Rat
Host Species Mouse
Regulatory Status RUO
Research Discipline Enzyme
Type Primary
Immunogen Purified rat liver estrogen sulfotransferase
Isotype IgG1
Purification Method Rat liver

Ensure accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Thermo Scientific™ Estrogen Sulfotransferase (Anti-Rat) Ab-1, Mouse Monoclonal Antibody.

Mammalian EST sulfurylates the hydroxyl group of estrogenic steroids by transferring the sulfate from a cosubstrate adenosine 3'-phosphate-5'-phosphosulfate. Sulfurylated steroids do not bind to the estrogen receptor with high affinity and, therefore, are hormonally inactive. EST plays an important role in controlling the intracellular level of the receptor-active estrogens.

Host Species: Mouse

Clone: EST.1

Isotype: IgG1

Species Reactivity: Rat. Others not known.

Epitope: Not determined

Immunogen: Purified rat liver estrogen sulfotransferase

Molecular Weight: ∽31 kDa

Positive Control: Rat liver

Cellular Localization: Cytoplasmic with some nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not verified)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO