Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Estrogen Sulfotransferase (Anti-Rat) Ab-1, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry and immunofluorescence experiments.

Manufacturer: thermo scientific™ lab vision™  MS233P1ABX

 View more versions of this product

Catalog No. MS233P1ABX

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Cross Reactivity Rat
Antigen Estrogen Sulfotransferase (Anti-Rat) Ab-1
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Quantity 100µL
Isotype IgG1
Format Concentrated
Regulatory Status RUO
Immunogen Purified rat liver estrogen sulfotransferase
Clone EST.1
Formulation Ab without BSA/Azide (Frozen)
Conjugate Unlabeled
Purification Method Rat liver
Type Primary
Research Discipline Enzyme
Classification Monoclonal
Concentration 1mg/mL

Ensure accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Thermo Scientific™ Estrogen Sulfotransferase (Anti-Rat) Ab-1, Mouse Monoclonal Antibody.

Mammalian EST sulfurylates the hydroxyl group of estrogenic steroids by transferring the sulfate from a cosubstrate adenosine 3'-phosphate-5'-phosphosulfate. Sulfurylated steroids do not bind to the estrogen receptor with high affinity and, therefore, are hormonally inactive. EST plays an important role in controlling the intracellular level of the receptor-active estrogens.

Host Species: Mouse

Clone: EST.1

Isotype: IgG1

Species Reactivity: Rat. Others not known.

Epitope: Not determined

Immunogen: Purified rat liver estrogen sulfotransferase

Molecular Weight: ∽31 kDa

Positive Control: Rat liver

Cellular Localization: Cytoplasmic with some nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not verified)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO